Invitrogen
Manufacturer Code:MA527665
Catalog # PIMA527665
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S395-68 |
Immunogen | Fusion protein amino acids 1-215 (full-length) of mouse Nav beta2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: neuronal voltage-gated sodium channel beta 2 subunit; Sodium channel beta 2; Sodium channel subunit beta-2; Sodium channel subunit beta-2 Scn2b UNQ326/PRO386 Gm183; sodium channel, voltage gated, type II beta subunit; sodium channel, voltage-gated, type II, beta polypeptide
Gene Aliases: 2810451E09Rik; AI840361; ATFB14; Gm183; SCN2B; SCNB2; UNQ326/PRO386
UniProt ID: (Human) O60939, (Rat) P54900, (Mouse) Q56A07
Entrez Gene ID: (Human) 6327, (Rat) 25349, (Mouse) 72821
Molecular Function: cell adhesion molecule ion channel sodium channel transporter voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.