Novus Biologicals
Manufacturer Code:NBP29324620UL
Catalog # NB028379
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 275-401 of human SCN2A (NP_001035232.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BFIC3 brain type 2 alpha subunit EIEE11 HBA HBSCI HBSCII Na(v)1.2 NAC2 Nav1.2 SCN2A1HBSC II SCN2A2 sodium channel voltage-gated type II alpha 1 polypeptide sodium channel voltage-gated type II alpha 2 polypeptide sodium channel voltage-gated type II alpha subunit voltage-gated sodium channel subtype II Voltage-gated sodium channel subunit alpha Nav1.2; HBSC II; Sodium channel protein brain II subunit alpha; Sodium channel protein type 2 subunit alpha; Sodium channel protein type II subunit alpha; sodium channel protein, brain type 2 alpha subunit; sodium channel, voltage gated, type II alpha subunit; sodium channel, voltage-gated, type II, alpha 1 polypeptide; sodium channel, voltage-gated, type II, alpha 2 polypeptide; sodium channel, voltage-gated, type II, alpha subunit; voltage-gated sodium channel subtype II; Voltage-gated sodium channel subunit alpha Nav1.2; voltage-gated sodium channel type II alpha subunit
Gene Aliases: BFIC3; BFIS3; BFNIS; EIEE11; HBA; HBSCI; HBSCII; Na(v)1.2; NAC2; Nav1.2; SCN2A; SCN2A1; SCN2A2
UniProt ID: (Human) Q99250
Entrez Gene ID: (Human) 6326
Molecular Function:
calcium channel
ion channel
sodium channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.