Invitrogen
Manufacturer Code:PA541003
Catalog # PIPA541003
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SCN1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: sodium channel beta 1-subunit; sodium channel beta-1 subunit; Sodium channel subunit beta-1; sodium channel voltage-gated type 1 beta polypeptide; sodium channel voltage-gated type I beta polypeptide; Sodium channel voltage-gated type I beta subunit GEFSP1; sodium channel, voltage gated, type I beta subunit; sodium channel, voltage-gated, type 1, beta polypeptide; sodium channel, voltage-gated, type I, beta; sodium channel, voltage-gated, type I, beta polypeptide; sodium channel, voltage-gated, type I, beta subunit
Gene Aliases: ATFB13; BRGDA5; GEFSP1; SCN1B
UniProt ID: (Human) Q07699, (Dog) Q4PPC4, (Bovine) Q17QN4, (Rabbit) P53788, (Mouse) P97952, (Rat) Q00954
Entrez Gene ID: (Human) 6324, (Horse) 100050415, (Dog) 484584, (Bovine) 618204, (Rabbit) 100009211, (Mouse) 20266, (Rat) 29686
Molecular Function:
ion channel
sodium channel
transporter
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.