Invitrogen
Manufacturer Code:PA524142
Catalog # PIPA524142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 63-90 amino acids from the N-terminal region of human SCN1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GEFSP1 sodium channel beta-1 subunit sodium channel subunit beta-1; Sodium channel subunit beta-1; sodium channel, voltage gated, type I beta subunit; sodium channel, voltage-gated, type I, beta; sodium channel, voltage-gated, type I, beta polypeptide
Gene Aliases: ATFB13; BRGDA5; GEFSP1; SCN1B
UniProt ID: (Human) Q07699, (Mouse) P97952
Entrez Gene ID: (Human) 6324, (Mouse) 20266
Molecular Function:
ion channel
sodium channel
transporter
voltage-gated ion channel
voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.