Invitrogen
Manufacturer Code:PA5113034
Catalog # PIPA5113034
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Sodium channel protein type 10 subunit alpha protein (992-1099AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FEPS2 hPN3 mPN3 Na(V)1.8 Nav1.8 Peripheral nerve sodium channel 3 PN3 SCN10A Sensory neuron sodium channel Sns Sodium channel protein type 10 subunit alpha Sodium channel protein type X subunit alpha sodium channel type X alpha polypeptide sodium channel voltage-gated type X alpha polypeptide sodium channel voltage gated type X alpha subunit sodium channel voltage-gated type 10 alpha polypeptide sodium channel voltage-gated type X alpha sodium channel voltage-gated type X alpha polypeptide sodium channel voltage-gated type X alpha subunit sodium voltage-gated channel alpha subunit 10 voltage gated sodium channel Voltage-gated sodium channel subunit alpha Nav1.8; Peripheral nerve sodium channel 3; PN3; Sodium channel protein type 10 subunit alpha; Sodium channel protein type X subunit alpha; sodium channel, voltage gated, type X alpha subunit; sodium channel, voltage-gated, type X, alpha polypeptide; sodium channel, voltage-gated, type X, alpha subunit; Voltage-gated sodium channel subunit alpha Nav1.8
Gene Aliases: FEPS2; hPN3; Nav1.8; PN3; SCN10A; SNS
UniProt ID: (Human) Q9Y5Y9
Entrez Gene ID: (Human) 6336
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.