Invitrogen
Manufacturer Code:PA5111804
Catalog # PIPA5111804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Guinea pig |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 1943-1956 of rat NaV1.8 (Intracellular C-terminus) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Guinea Pig Polyclonal Antibody
For Research Use Only
Protein Aliases: FEPS2 hPN3 mPN3 Na(V)1.8 Nav1.8 Peripheral nerve sodium channel 3 PN3 SCN10A Sensory neuron sodium channel Sns Sodium channel protein type 10 subunit alpha Sodium channel protein type X subunit alpha Voltage-gated sodium channel subunit alpha Nav1.8; Peripheral nerve sodium channel 3; PN3; Sensory neuron sodium channel; Sodium channel protein type 10 subunit alpha; Sodium channel protein type X subunit alpha; sodium channel type X alpha polypeptide; sodium channel voltage-gated type X alpha polypeptide; sodium channel, voltage-gated, type 10, alpha polypeptide; Voltage-gated sodium channel subunit alpha Nav1.8
Gene Aliases: Na(V)1.8; PN3; Scn10a; Sns
UniProt ID: (Rat) Q62968
Entrez Gene ID: (Rat) 29571
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.