Novus Biologicals
Manufacturer Code:NBP222349PEATT594
Catalog # N222349P594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NVL-07 |
Immunogen | Recombinant intracellular part of human SCIMP. |
Conjugate | PE/ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: DTFT5783; SLP adapter and CSK-interacting membrane protein; SLP65/SLP76, Csk-interacting membrane protein; transmembrane protein C17orf87; transmembrane protein C17orf87 C17orf87 chromosome 17 open reading frame 87 DTFT5783 FLJ32580 SLP adapter and CSK-interacting membrane protein SLP adaptor and CSK interacting membrane protein SLP65/SLP76 Csk-interacting membrane protein UNQ5783
Gene Aliases: C17orf87; SCIMP; UNQ5783; UNQ5783/PRO16090
UniProt ID: (Human) Q6UWF3
Entrez Gene ID: (Human) 388325
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.