Invitrogen
Manufacturer Code:PA569992
Catalog # PIPA569992
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SCHIP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: neurofibromatosis 2 interacting protein; SCHIP-1; schwannomin interacting protein 1; Schwannomin-interacting protein 1; Schwannomin-interacting protein 1 SCHIP-1 SCHIP1
Gene Aliases: Nf2ip; SCHIP-1; SCHIP1; zgc:110585
UniProt ID: (Human) P0DPB3, (Mouse) P0DPB4
Entrez Gene ID: (Human) 29970, (Bovine) 535716, (Mouse) 30953, (Rat) 295105, (Zebrafish) 550605
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.