OriGene
Manufacturer Code:TA808451
Catalog # 50-168-2143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI10D10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 66-190 of human SCFD1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C14orf163 RA410 SLY1 SLY1P STXBP1L2; Sec1 family domain-containing protein 1; SLY1 homolog; Sly1p; Syntaxin-binding protein 1-like 2; vesicle transport-related protein
Gene Aliases: C14orf163; FKSG23; KIAA0917; RA410; SCFD1; SLY1; SLY1P; STXBP1L2
UniProt ID: (Human) Q8WVM8
Entrez Gene ID: (Human) 23256
Molecular Function: membrane traffic protein membrane trafficking regulatory protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.