Novus Biologicals
Manufacturer Code:NBP177030
Catalog # NBP177030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against an 18 amino acid synthetic peptide from near the center of human SCF. The immunogen is located within amino acids 100 - 150 of SCF. Amino Acid Squence: KENSSKDLKKSFKSPEPR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-Kit ligand; DKFZp686F2250 familial progressive hyperpigmentation 2 FPH2 KIT ligand Kitl KL-1 Mast cell growth factor MGFSHEP7 SCFStem cell factor SFc-Kit ligand steel factor; familial progressive hyperpigmentation 2; Kit ligand; Mast cell growth factor; MGF; SCF; sKITLG; Soluble KIT ligand; steel factor; Stem cell factor
Gene Aliases: DCUA; DFNA69; FPH2; FPHH; Kitl; KITLG; KL-1; MGF; SCF; SF; SHEP7
UniProt ID: (Human) P21583
Entrez Gene ID: (Human) 4254
Molecular Function:
cell adhesion molecule
growth factor
membrane-bound signaling molecule
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.