Novus Biologicals
Manufacturer Code:NBP294369100UL
Catalog # NB023291
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-75 of human SCD5 (NP_001032671.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACOD4Acyl-CoA-desaturase 4 FADS4 FLJ21032 HSCD5Stearoyl-CoA 9-desaturase SCD2 SCD4EC 1.14.19.1 stearoyl-CoA desaturase 4 stearoyl-CoA desaturase 5; Acyl-CoA-desaturase 4; HSCD5; Stearoyl-CoA 9-desaturase; Stearoyl-CoA desaturase 2; stearoyl-CoA desaturase 4; Stearoyl-CoA desaturase 5
Gene Aliases: ACOD4; FADS4; HSCD5; SCD2; SCD4; SCD5
UniProt ID: (Human) Q86SK9
Entrez Gene ID: (Human) 79966
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.