Abnova
Manufacturer Code:PAB15990
Catalog # 89-118-000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of <i>Arabidopsis thaliana</i> SCD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SCD1.
For Research Use Only
Protein Aliases: DENN domain and WD repeat-containing protein SCD1; F27J15.17; F27J15_17; Protein STOMATAL CYTOKINESIS DEFECTIVE 1; SCD1 protein; STOMATAL CYTOKINESIS-DEFECTIVE 1
Gene Aliases: AT1G49040; F27J15.16/F27J15.17; F27J15.17; F27J15_17; SCD1; STOMATAL CYTOKINESIS-DEFECTIVE 1
UniProt ID: (Arabidopsis) Q8RXA7
Entrez Gene ID: (Arabidopsis) 841327
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.