Invitrogen
Manufacturer Code:MA514885
Catalog # PIMA514885
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | G.545.10 |
Immunogen | Synthetic peptide corresponding to residues surrounding Leu35 of mouse SCD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA desaturase 1; asebia; Delta(9)-desaturase 1; Delta-9 desaturase 1; Fatty acid desaturase 1; OTTHUMP00000020280 acyl-CoA desaturase delta(9)-desaturase delta-9-desaturase fatty acid desaturase predicted protein of HQ0998 stearoyl-CoA desaturase opposite strand; Stearoyl-CoA desaturase 1
Gene Aliases: AA589638; ab; AI265570; Scd; Scd-1; Scd1
UniProt ID: (Mouse) P13516
Entrez Gene ID: (Mouse) 20249
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.