Invitrogen
Manufacturer Code:MA527542
Catalog # PIMA527542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CD.E10 |
Immunogen | Full length human recombit protein against SCD1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA desaturase; Acyl-CoA desaturase 1; asebia; Delta(9)-desaturase; Delta(9)-desaturase 1; Delta-9 desaturase; Delta-9 desaturase 1; delta-9-desaturase; Fatty acid desaturase; Fatty acid desaturase 1; hSCD1; predicted protein of HQ0998; Stearoyl-CoA desaturase; stearoyl-CoA desaturase (delta-9-desaturase); Stearoyl-CoA desaturase 1; stearoyl-CoA desaturase opposite strand; Stearoyl-CoA desaturase-1 SCD1 CD.E10
Gene Aliases: AA589638; ab; AI265570; FADS5; hSCD1; MSTP008; SCD; Scd-1; SCD1; SCDOS
UniProt ID: (Human) D3DR68, (Mouse) P13516, (Rat) P07308
Entrez Gene ID: (Human) 6319, (Mouse) 20249, (Rat) 246074
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.