Abnova
Manufacturer Code:H00006319M02A
Catalog # 89-105-404
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B7 |
Immunogen | SCD (NP_005054.31 a.a. &bsim 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant SCD.
For Research Use Only
Protein Aliases: Acyl-CoA desaturase; Delta(9)-desaturase; Delta-9 desaturase; delta-9-desaturase; FADS5; Fatty acid desaturase; hSCD1; MSTP008; predicted protein of HQ0998; SCD1; Stearoyl-CoA desaturase; stearoyl-CoA desaturase (delta-9-desaturase); stearoyl-CoA desaturase opposite strand
Gene Aliases: FADS5; hSCD1; MSTP008; SCD; SCD1; SCDOS
UniProt ID: (Human) D3DR68
Entrez Gene ID: (Human) 6319
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.