Invitrogen
Manufacturer Code:PA522996
Catalog # PIPA522996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of the human protein (within residues 50-100). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acyl-CoA desaturase; Delta(9)-desaturase; Delta-9 desaturase; delta-9-desaturase; Fatty acid desaturase; hSCD1; predicted protein of HQ0998; PRO1933 FADS5 MSTP008 SCD1 SCDOS acyl-CoA desaturase delta(9)-desaturase delta-9 desaturase delta-9-desaturase fatty acid desaturase predicted protein of HQ0998 stearoyl-CoA desaturase opposite strand; Stearoyl-CoA desaturase; stearoyl-CoA desaturase (delta-9-desaturase); stearoyl-CoA desaturase opposite strand
Gene Aliases: FADS5; hSCD1; MSTP008; SCD; SCD1; SCDOS
UniProt ID: (Human) D3DR68
Entrez Gene ID: (Human) 6319
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.