Invitrogen
Manufacturer Code:PA569978
Catalog # PIPA569978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SCCPDH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NET11 RP11-439E19.2 probable saccharopine dehydrogenase saccharopine dehydrogenase-like oxidoreductase; probable saccharopine dehydrogenase; Saccharopine dehydrogenase-like oxidoreductase
Gene Aliases: AW214504; C330023F11Rik; CGI-49; NET11; RGD1311440; SCCPDH
UniProt ID: (Human) Q8NBX0, (Bovine) Q3T067, (Mouse) Q8R127, (Rat) Q6AY30
Entrez Gene ID: (Human) 51097, (Bovine) 507289, (Rabbit) 100346223, (Horse) 100058947, (Mouse) 109232, (Rat) 305021, (Guinea pig) 100731395
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.