Invitrogen
Manufacturer Code:PA5102589
Catalog # PIPA5102589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SCCPDH corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW214504 C330023F11Rik CGI 49 CGI49 CGI-49 NET11 probable saccharopine dehydrogenase RGD1311440 RP11-439E19.2 saccharopine dehydrogenase (putative) saccharopine dehydrogenase-like oxidoreductase SCCPDH SCPDH; probable saccharopine dehydrogenase; Saccharopine dehydrogenase-like oxidoreductase
Gene Aliases: AW214504; C330023F11Rik; CGI-49; NET11; RGD1311440; SCCPDH
UniProt ID: (Human) Q8NBX0, (Mouse) Q8R127, (Rat) Q6AY30
Entrez Gene ID: (Human) 51097, (Mouse) 109232, (Rat) 305021
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.