Novus Biologicals
Manufacturer Code:NBP297630AF700
Catalog # NB097615
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SCCPDH fragment. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI-49 EC 1.5.1.9 FLJ43187 NET11 probable saccharopine dehydrogenase RP11-439E19.2 saccharopine dehydrogenase (putative); probable saccharopine dehydrogenase; Saccharopine dehydrogenase-like oxidoreductase
Gene Aliases: CGI-49; NET11; SCCPDH
UniProt ID: (Human) Q8NBX0
Entrez Gene ID: (Human) 51097
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.