Proteintech
Manufacturer Code:249411AP150UL
Catalog # 50-173-4632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SCCPDH Fusion Protein Ag11551 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI 49 NET11 RP11 439E19.2 SCCPDH; probable saccharopine dehydrogenase; Saccharopine dehydrogenase-like oxidoreductase
Gene Aliases: AW214504; C330023F11Rik; CGI-49; NET11; SCCPDH
UniProt ID: (Human) Q8NBX0, (Mouse) Q8R127
Entrez Gene ID: (Human) 51097, (Mouse) 109232
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.