Abnova
Manufacturer Code:PAB12597
Catalog # 89-115-933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Blocking Assay (BLOCK) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Scarb1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant Scarb1.
For Research Use Only
Protein Aliases: AI120173; CD36; Cd36l1; Cla-1; Cla1; D5Ertd460e; HDL QTL 1; mSR-BI; Scavenger receptor class B member 1; scavenger receptor class B type I; scavenger receptor class B1; SR-B1; SR-BI; SRB1; SRBI
Gene Aliases: AI120173; CD36; Cd36l1; Cla-1; Cla1; D5Ertd460e; Hdlq1; Hlb398; mSR-BI; Scarb1; SR-B1; SR-BI; Srb1; SRBI
UniProt ID: (Mouse) Q61009
Entrez Gene ID: (Mouse) 20778
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.