R&D Systems
Manufacturer Code:FAB4900N100UG
Catalog # FAB4900N100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 673527 |
Immunogen | NS0 mouse myeloma cell line transfected with human SCARA5 Arg83-His495 Accession # Q6ZMJ2 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: FLJ23907 MGC45780 scavenger receptor class A member 5 (putative) Tesr testis expressed scavenger receptor UNQ2938/PRO28700; Scavenger receptor class A member 5; scavenger receptor class A, member 5 (putative); Scavenger receptor hlg; testis expressed scavenger receptor
Gene Aliases: NET33; SCARA5; Tesr; UNQ2938/PRO28700
UniProt ID: (Human) Q6ZMJ2
Entrez Gene ID: (Human) 286133
Molecular Function:
hydrolase
oxidase
oxidoreductase
protease
receptor
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.