Invitrogen
Manufacturer Code:PA520766
Catalog # PIPA520766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide near the carboxy terminus of human SCARA5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Scavenger receptor class A member 5; scavenger receptor class A, member 5 (putative); Scavenger receptor hlg; testis expressed scavenger receptor; UNQ2938/PRO28700 NET33 Tesr scavenger receptor class A member 5 scavenger receptor hlg testis expressed scavenger receptor
Gene Aliases: NET33; SCARA5; Tesr; UNQ2938/PRO28700
UniProt ID: (Human) Q6ZMJ2
Entrez Gene ID: (Human) 286133
Molecular Function:
hydrolase
oxidase
oxidoreductase
protease
receptor
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.