Invitrogen
Manufacturer Code:PA528982
Catalog # PIPA528982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 977 and 1224 of Human SREBF chaperone protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SCAP; Sreb cleavage-activating protein; SREBP cleavage activating protein; SREBP cleavage-activating protein; SREBP cleavage-activating protein sterol regulatory element binding protein cleavage-activating protein sterol regulatory element-binding protein cleavage-activating protein; sterol regulatory element binding protein cleavage-activating protein; Sterol regulatory element-binding protein cleavage-activating protein
Gene Aliases: 9530044G19; KIAA0199; mKIAA0199; PSEC0227; SCAP
UniProt ID: (Human) Q12770, (Mouse) Q6GQT6
Entrez Gene ID: (Human) 22937, (Mouse) 235623
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.