Novus Biologicals
Manufacturer Code:NBP204113AF405
Catalog # NBP2043AFY
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 450 and 500 of human Sterol Regulatory Element Binding Protein Ceavage-Activating Protein using the numbering given in entry NP_036367.2 (GeneID 22937). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0199 PSEC0227 SREBF chaperone SREBP cleavage-activating protein sterol regulatory element binding protein cleavage-activating protein; SCAP; SREBP cleavage-activating protein; sterol regulatory element binding protein cleavage-activating protein; Sterol regulatory element-binding protein cleavage-activating protein
Gene Aliases: KIAA0199; PSEC0227; SCAP
UniProt ID: (Human) Q12770
Entrez Gene ID: (Human) 22937
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.