Invitrogen
Manufacturer Code:PA552408
Catalog # PIPA552408
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SCAND3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ1186N24.3 (novel zinc finger protein); SCAN domain containing 3; SCAN domain-containing protein 3; Transposon-derived Buster4 transposase-like protein; Zinc finger BED domain-containing protein 9; zinc finger protein 305 pseudogene 2; zinc finger protein 452; zinc finger, BED-type containing 9
Gene Aliases: Buster4; dJ1186N24.3; KIAA1925; SCAND3; ZBED9; ZFP38-L; ZNF305P2; ZNF452
UniProt ID: (Human) Q8TCN2
Entrez Gene ID: (Human) 114821
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.