Abnova
Manufacturer Code:H00006309K
Catalog # 89-202-432
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SC5DL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SC5DL peptide using ARM Technology.
For Research Use Only
Protein Aliases: 3beta-hydroxysteroid-delta5-desaturase; C-5 sterol desaturase; Delta(7)-sterol 5-desaturase; Delta(7)-sterol C5(6)-desaturase; ERG3 S5DES SC5D; fungal ERG3, delta-5-desaturase-like; Lathosterol 5-desaturase; lathosterol dehydrogenase; Lathosterol oxidase; Sterol-C5-desaturase; sterol-C5-desaturase (ERG3 delta-5-desaturase homolog, S. cerevisiae)-like
Gene Aliases: ERG3; S5DES; SC5D; SC5DL
UniProt ID: (Human) O75845
Entrez Gene ID: (Human) 6309
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.