Invitrogen
Manufacturer Code:PA518580
Catalog # PIPA518580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (KERQERKQRLQEN) corresponding to the internal amino acids of SECISBP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: SBP2 RP11-89K14.1 SECISBP2 SECIS binding protein 2 DKFZp686C09169 OTTHUMP00000064929 OTTHUMP00000064930 OTTHUMP00000064931 OTTHUMP00000064932 selenocysteine insertion sequence binding protein 2; SECIS-binding protein 2; Selenocysteine insertion sequence-binding protein 2
Gene Aliases: 2210413N07Rik; 2810012K13Rik; SBP2; SECISBP2
UniProt ID: (Human) Q7L1Z0
Entrez Gene ID: (Human) 79048, (Mouse) 75420
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.