Proteintech
Manufacturer Code:176181AP150UL
Catalog # 50-173-1686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SBDS Fusion Protein Ag11814 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI 97 SBDS SDS SWDS; Protein 22A3; Ribosome maturation protein SBDS; Shwachman-Bodian-Diamond syndrome; Shwachman-Bodian-Diamond syndrome homolog; Shwachman-Bodian-Diamond syndrome protein; Shwachman-Bodian-Diamond syndrome protein homolog
Gene Aliases: 4733401P19Rik; AI836084; CGI-97; SBDS; SDS; SWDS
UniProt ID: (Human) Q9Y3A5, (Mouse) P70122, (Rat) Q5RK30
Entrez Gene ID: (Human) 51119, (Mouse) 66711, (Rat) 288615
Molecular Function:
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.