Novus Biologicals
Manufacturer Code:NBP273990APC
Catalog # NB273990APC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C9 |
Immunogen | Full length recombinant protein of human SAT2 (NP_597998) produced in HEK293T cells. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Diamine acetyltransferase 2; diamine N-acetyltransferase 2; diamine N-acetyltransferase 2 EC 2.3.1.57 Polyamine N-acetyltransferase 2 S Spermidine/spermine N(1)-acetyltransferase 2 spermidine/spermine N1-acetyltransferase 2 spermidine/spermine N1-acetyltransferase family member 2 SSAT2diamine acetyltransferase 2 Thialysine N-epsilon-acetyltransferase; polyamine N-acetyltransferase 2; Spermidine/spermine N(1)-acetyltransferase 2; SSAT-2; Thialysine N-epsilon-acetyltransferase
Gene Aliases: SAT2; SSAT2
UniProt ID: (Human) Q96F10
Entrez Gene ID: (Human) 112483
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.