Proteintech
Manufacturer Code:162461AP150UL
Catalog # 50-173-0825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SAT2 Fusion Protein Ag9179 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Diamine acetyltransferase 2; Diamine acetyltransferase 2 SAT2 SSAT2; diamine N-acetyltransferase 2; polyamine N-acetyltransferase 2; Spermidine/spermine N(1)-acetyltransferase 2; spermidine/spermine N1-acetyl transferase 2; SSAT-2; Thialysine N-epsilon-acetyltransferase
Gene Aliases: 2610016A03Rik; SAT2; SSAT-2; SSAT2
UniProt ID: (Human) Q96F10, (Mouse) Q6P8J2
Entrez Gene ID: (Human) 112483, (Rat) 360547, (Mouse) 69215
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.