Invitrogen
Manufacturer Code:PA5106434
Catalog # PIPA5106434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SAT1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA617398 Ab2-402 dc21 Diamine acetyltransferase 1 diamine acetyltransferase 1 spermidine/spermine N1-acetyltransferase 1 diamine N-acetyltransferase 1 diamine N-acetyltransferase 1 a EC 2.3.1.57 epid34 hypothetical protein LOC508861 kfsd KFSDX Polyamine N-acetyltransferase 1 putrescine acetyltransferase SAT SAT1 sat1.S sat1a sat1-a sat1-b Spermidine / spermine N1-acyltransferase (diamine acetyltransferase) spermidine/spermine N(1)-acetyltransferase 1 spermidine/spermine N1-acetyl transferase 1 spermidine/spermine N1-acetyltransferase 1 spermidine/spermine N1-acetyltransferase 1 a spermidine/spermine N1-acetyltransferase 1 S homeolog spermidine/spermine N1-acetyltransferase alpha spermidine/spermine N-acetyltransferase SSAT SSAT-1 XELAEV_18014544mg; Diamine acetyltransferase 1; diamine N-acetyltransferase 1; Polyamine N-acetyltransferase 1; Putrescine acetyltransferase; Spermidine / spermine N1-acyltransferase (diamine acetyltransferase); Spermidine/spermine N(1)-acetyltransferase 1; spermidine/spermine N1-acetyltransferase alpha; SSAT; SSAT-1
Gene Aliases: AA617398; Ab2-402; DC21; KFSD; KFSDX; SAT; SAT1; SSAT; SSAT-1
UniProt ID: (Human) P21673, (Mouse) P48026
Entrez Gene ID: (Human) 6303, (Mouse) 20229, (Rat) 302642
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.