Abnova
Manufacturer Code:PAB12044
Catalog # 89-115-552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 100-171 of human SAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for short term. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SAT1.
For Research Use Only
Protein Aliases: DC21; Diamine acetyltransferase 1; diamine N-acetyltransferase 1; KFSD; Polyamine N-acetyltransferase 1; Putrescine acetyltransferase; SAT; Spermidine/spermine N(1)-acetyltransferase 1; spermidine/spermine N1-acetyltransferase alpha; SSAT; SSAT-1
Gene Aliases: DC21; KFSD; KFSDX; SAT; SAT1; SSAT; SSAT-1
UniProt ID: (Human) P21673
Entrez Gene ID: (Human) 6303
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.