Proteintech
Manufacturer Code:266111AP150UL
Catalog # 50-173-5398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SAT1 Fusion Protein Ag24306 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EDM4 SAT 1 SAT1 SLC26A1 Sulfate anion transporter 1; SAT-1; solute carrier family 26 (anion exchanger), member 1; solute carrier family 26 (sulfate transporter), member 1; Solute carrier family 26 member 1; solute carrier family 26, member 1; sulfate anion tranporter AT1; Sulfate anion transporter 1; sulfate anion transporter-1; sulfate/anion transporter SAT-1 protein
Gene Aliases: CAON; EDM4; SAT-1; SAT1; SLC26A1
UniProt ID: (Human) Q9H2B4, (Mouse) P58735
Entrez Gene ID: (Human) 10861, (Mouse) 231583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.