Invitrogen
Manufacturer Code:PA116992
Catalog # 01-670-145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues 100-171 of human SAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Diamine acetyltransferase 1; Diamine acetytransferase 1 EC 2.3.1.57 Spermidine/spermine N(1)-acetyltransferase 1 SSAT SSAT-1 Puterscine acetyltransferase Polyamine N-acetylt; diamine N-acetyltransferase 1; Polyamine N-acetyltransferase 1; Putrescine acetyltransferase; SAT; Spermidine / spermine N1-acyltransferase (diamine acetyltransferase); Spermidine/spermine N(1)-acetyltransferase 1; spermidine/spermine N-acetyltransferase; spermidine/spermine N1-acetyltransferase 1 a; spermidine/spermine N1-acetyltransferase alpha; SSAT; SSAT-1
Gene Aliases: AA617398; Ab2-402; DC21; epid34; KFSD; KFSDX; SAT; SAT1; sat1-a; sat1-b; sat1a; SSAT; SSAT-1; XELAEV_18014544mg
UniProt ID: (Chicken) Q8AXL1, (Bovine) Q3T0Q0, (Human) P21673, (Pig) Q28999, (Mouse) P48026
Entrez Gene ID: (Chicken) 374006, (Bovine) 508861, (Human) 6303, (Pig) 397645, (Goat) 100860804, (Mouse) 20229, (Rat) 302642, (Xenopus) 100036974
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.