Invitrogen
Manufacturer Code:PA570305
Catalog # PIPA570305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human SASH3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Novel SH3 and SAM domain containing protein; SAM and SH3 domain-containing protein 3; SH3 protein expressed in lymphocytes; SH3 protein expressed in lymphocytes homolog
Gene Aliases: 1200013B08Rik; 753P9; AW413946; CXorf9; HACS2; HXCXORF9; RGD1560293; SASH3; SH3D6C; SLY; SLY1
UniProt ID: (Human) O75995, (Bovine) A0JN71, (Mouse) Q8K352
Entrez Gene ID: (Human) 54440, (Bovine) 534137, (Dog) 492125, (Horse) 100054731, (Rabbit) 100347012, (Rat) 317578, (Guinea pig) 100732342, (Mouse) 74131
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.