Abnova
Manufacturer Code:PAB20031
Catalog # 89-121-532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SASH3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SASH3.
For Research Use Only
Protein Aliases: 753P9; CXorf9; HACS2; SAM and SH3 domain-containing protein 3; SH3 protein expressed in lymphocytes homolog; SH3D6C; SLY
Gene Aliases: 753P9; CXorf9; HACS2; SASH3; SH3D6C; SLY
UniProt ID: (Human) O75995
Entrez Gene ID: (Human) 54440
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.