Novus Biologicals
Manufacturer Code:NBP126651
Catalog # NBP126651
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human SAM and SH3 domain containing 1 using the numbering given in entry NP_056093.3 (GeneID 23328). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2500002E12Rik; dJ323M4 dJ323M4.1 KIAA0790RP3-323M4.1 PEPE1 Proline-glutamate repeat-containing protein SAM and SH3 domain containing 12500002E12Rik SAM and SH3 domain-containing protein 1 SH3D6A; Proline-glutamate repeat-containing protein; SAM and SH3 domain-containing protein 1
Gene Aliases: dJ323M4; dJ323M4.1; KIAA0790; PEPE1; SASH1; SH3D6A
UniProt ID: (Human) O94885
Entrez Gene ID: (Human) 23328
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.