Invitrogen
Manufacturer Code:PA518120
Catalog # PIPA518120
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (MSNADFAKLFLRK) corresponding to the C-terminus amino acids of SART3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: HIV-1 Tat-interacting protein of 110kDa; hSART-3; p110 nuclear RNA-binding protein; SART-3; SART3 squamous cell carcinoma antigen recognised by T cells 3 TIP110 KIAA0156 p110(nrb) RP11-13G14 tat-interacting protein of 110 kDa DSAP1 MGC138188 P100 p110; squamous cell carcinoma antigen recognized by T cells 3; Squamous cell carcinoma antigen recognized by T-cells 3; Tat-interacting protein of 110 kDa; Tip110
Gene Aliases: DSAP1; KIAA0156; P100; p110; p110(nrb); RP11-13G14; SART3; TIP110
UniProt ID: (Human) Q15020
Entrez Gene ID: (Human) 9733
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.