Invitrogen
Manufacturer Code:PA5103326
Catalog # PIPA5103326
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SART3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU045857 DSAP1 HIV-1 Tat-interacting protein of 110kDa hSART-3 Kiaa0156 MGC138188 mKIAA0156 mSART-3 P100 p110 p110 nuclear RNA-binding protein p110(nrb) RP11-13G14 SART3 SART-3 squamous cell carcinoma antigen recognized by T cells 3 squamous cell carcinoma antigen recognized by T-cells 3 Tat-interacting protein of 110 kDa Tip110 Tumor-rejection antigen SART3; HIV-1 Tat-interacting protein of 110kDa; hSART-3; mSART-3; p110 nuclear RNA-binding protein; SART-3; squamous cell carcinoma antigen recognized by T cells 3; Squamous cell carcinoma antigen recognized by T-cells 3; Tat-interacting protein of 110 kDa; Tip110; Tumor-rejection antigen SART3
Gene Aliases: AU045857; DSAP1; KIAA0156; mKIAA0156; P100; p110; p110(nrb); RP11-13G14; SART3; TIP110
UniProt ID: (Human) Q15020, (Mouse) Q9JLI8
Entrez Gene ID: (Human) 9733, (Mouse) 53890
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.