Novus Biologicals
Manufacturer Code:NB1001893
Catalog # NB1001893
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-MSNADFAKLFLRK corresponding to C-Terminus according to NP_055521.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DSAP1 hSART-3 KIAA0156squamous cell carcinoma antigen recognised by T cells 3 P100 p110 p110(nrb) RP11-13G14 SART-3 squamous cell carcinoma antigen recognized by T cells 3 squamous cell carcinoma antigen recognized by T-cells 3 Tat-interacting protein of 110 kDa Tip110 TIP110MGC138188; HIV-1 Tat-interacting protein of 110kDa; hSART-3; p110 nuclear RNA-binding protein; SART-3; squamous cell carcinoma antigen recognized by T cells 3; Squamous cell carcinoma antigen recognized by T-cells 3; Tat-interacting protein of 110 kDa; Tip110
Gene Aliases: DSAP1; KIAA0156; P100; p110; p110(nrb); RP11-13G14; SART3; TIP110
UniProt ID: (Human) Q15020
Entrez Gene ID: (Human) 9733
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.