Invitrogen
Manufacturer Code:PA5111017
Catalog # PIPA5111017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human SART1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ara1 Haf Hom s 1 HOMS1 hSART-1 hSnu66 Hypoxia-associated factor MGC2038 Sart1 SART-1 SART1259 SNRNP110 Snu66 SNU66 homolog Squamous cell carcinoma antigen recognized by T-cells 1 U4/U6.U5 tri-snRNP-associated 110 kDa protein U4/U6.U5 tri-snRNP-associated protein 1 U5-110K; Hom s 1; hSART-1; hSnu66; Hypoxia-associated factor; IgE autoantigen; mSART-1; rSART-1; SART-1; SART1(259) protein; SART1(800) protein; small nuclear ribonucleoprotein 110kDa (U4/U6.U5); SNU66 homolog; squamous cell carcinoma antigen recognised by T cells; squamous cell carcinoma antigen recognized by T cells 1; Squamous cell carcinoma antigen recognized by T-cells 1; U4/U6.U5 tri-snRNP-associated 110 kDa protein; U4/U6.U5 tri-snRNP-associated protein 1
Gene Aliases: Ara1; Haf; HOMS1; SART1; SART1259; SNRNP110; Snu66; U5-110K
UniProt ID: (Human) O43290, (Mouse) Q9Z315, (Rat) Q5XIW8
Entrez Gene ID: (Human) 9092, (Mouse) 20227, (Rat) 29678
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.