Abnova
Manufacturer Code:H00009092K
Catalog # 89-203-021
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SART1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human SART1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Ara1 HOMS1 MGC2038 SART1259 SNRNP110 Snu66; Hom s 1; hSART-1; hSnu66; IgE autoantigen; SART-1; SART1(259) protein; SART1(800) protein; small nuclear ribonucleoprotein 110kDa (U4/U6.U5); SNU66 homolog; squamous cell carcinoma antigen recognised by T cells; squamous cell carcinoma antigen recognized by T cells 1; Squamous cell carcinoma antigen recognized by T-cells 1; U4/U6.U5 tri-snRNP-associated 110 kDa protein; U4/U6.U5 tri-snRNP-associated protein 1
Gene Aliases: Ara1; HOMS1; SART1; SART1259; SNRNP110; Snu66
UniProt ID: (Human) O43290
Entrez Gene ID: (Human) 9092
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.