Invitrogen
Manufacturer Code:PA531473
Catalog # PIPA531473
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 315 of Human SARS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mitochondrial seryl-tRNA synthetase; SARS SARSM SERS SYS SerRS SerRSmt mtSerRS mitochondrial seryl-tRNA synthetase serine tRNA ligase 2 mitochondrial serine--tRNA ligase mitochondrial serine-tRNA ligase mitochondrial seryl-tRNA synthetase mitochondrial seryl-tRNA(Ser/Sec) synthetase; serine tRNA ligase 2, mitochondrial; Serine--tRNA ligase, mitochondrial; serine-tRNA ligase, mitochondrial; SerRS; SerRSmt; Seryl-tRNA synthetase; seryl-tRNA synthetase, mitochondrial; Seryl-tRNA(Ser/Sec) synthetase
Gene Aliases: mtSerRS; SARS; SARS2; SARSM; SerRS; SerRSmt; SERS; SYS
UniProt ID: (Human) Q9NP81
Entrez Gene ID: (Human) 54938
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.