Invitrogen
Manufacturer Code:PA514984
Catalog # PIPA514984
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 160-188 amino acids from the N-terminal region of human SARS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mitochondrial seryl-tRNA synthetase; serine tRNA ligase 2, mitochondrial; Serine--tRNA ligase, mitochondrial; serine-tRNA ligase, mitochondrial; SerRS; SerRSmt; Seryl-tRNA synthetase; Seryl-tRNA synthetase mitochondrial &lsqbPrecursor] EC 6.1.1.11 Seryl-tRNA(Ser/Sec) synthetase Serine--tRNA ligase SerRSmt SARS2 SARSM; seryl-tRNA synthetase, mitochondrial; Seryl-tRNA(Ser/Sec) synthetase
Gene Aliases: mtSerRS; SARS; SARS2; SARSM; SerRS; SerRSmt; SERS; SYS
UniProt ID: (Human) Q9NP81
Entrez Gene ID: (Human) 54938
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.