Novus Biologicals
Manufacturer Code:NB10056683SS
Catalog # NB10056683T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
In Situ Hybridization (ISH) (ISH) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 354-370 (NKHIDAYKTFPPTEPKK-C) from the N (SARS Nucleocapsid) for the Human SARS coronavirus (Genbank accession no. YP_009724397.2) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COVID-19 nucleocapsid protein N N protein N structural protein NC Nucleocapsid protein Nucleoprotein Protein N SARS coronavirus N protein SARS coronavirus nucleocapsid protein SARS CoV SARS CoV N protein SARS CoV nucleocapsid protein SARS N protein SARS Nucleoprotein SARSCoV SARSCoV N protein SARSCoV nucleocapsid protein SARSCOV2 N protein SARS-COV-2 nucleocapsid protein SARS-COV-2 Nucleoprotein Severe acute respiratory syndrome
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.