Proteintech
Manufacturer Code:222921AP150UL
Catalog # 50-173-3819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310075M17Rik; ANDD CMRD GTBPB GTP binding protein B GTP binding protein SAR1b SAR1 homolog B (S. cerevisiae) SAR1B SARA2 SARB; GTBPB; GTP-binding protein B; GTP-binding protein SAR1b; GTP-binding protein Sara; SAR1 gene homolog B; SAR1 homolog B; SAR1a gene homolog 2; SAR1b gene homolog; secretion associated, Ras related GTPase 1B
Gene Aliases: 2310075M17Rik; 2900019I22Rik; ANDD; CMRD; GTBPB; SAR1B; SARA1B; SARA2; SARB
UniProt ID: (Human) Q9Y6B6, (Rat) Q5HZY2, (Mouse) Q9CQC9
Entrez Gene ID: (Human) 51128, (Rat) 287276, (Mouse) 66397
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.