Proteintech
Manufacturer Code:222911AP150UL
Catalog # 50-173-3818
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COPII associated small GTPase GTP binding protein SAR1a masra2 SAR1 SAR1 homolog A (S. cerevisiae) SAR1A Sara SARA1; COPII-associated small GTPase; GTP-binding protein SAR1a; SAR1 gene homolog A; SAR1 homolog A; SAR1a gene homolog 1; secretion associated, Ras related GTPase 1A
Gene Aliases: 1600019H17Rik; masra2; SAR1; SAR1A; SARA; SARA1
UniProt ID: (Human) Q9NR31
Entrez Gene ID: (Human) 56681, (Mouse) 20224, (Rat) 361842
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.