Invitrogen
Manufacturer Code:PA585827
Catalog # PIPA585827
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus region of mouse SAP97. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B130052P05Rik antibody Dlg1 antibody Dlgh1 antibody E dlg/SAP97 antibody discs large homolog 1 (Drosophila) antibody mKIAA4187 antibody; Disks large homolog 1; Drosophila discs-large tumor suppressor homologue (synapse associated protein); E-dlg/SAP97; Embryo-dlg/synapse-associated protein 97; SAP-97; Synapse-associated protein 97
Gene Aliases: B130052P05Rik; Dlg1; Dlgh1; E-dlg/SAP97; mKIAA4187; SAP-97; SAP97
UniProt ID: (Rat) Q62696, (Mouse) Q811D0
Entrez Gene ID: (Rat) 25252, (Mouse) 13383
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.