Invitrogen
Manufacturer Code:PA518790
Catalog # PIPA518790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (NSKTRDKGEIPDD) corresponding to the internal amino acids of DLG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: discs large homolog 1; discs large homolog 1, scribble cell polarity complex component; discs, large homolog 1; discs, large homolog 1 (Drosophila) transcript variant 6-v1; discs, large homolog 1 (Drosophila) transcript variant 6-v2; Disks large homolog 1; DLG1 discs large homolog 1 (Drosophila) DKFZp761P0818 DKFZp781B0426 DLGH1 SAP97 dJ1061C18.1.1 hdlg discs large homolog 1 discs large homolog 1 presynaptic protein SAP97 synapse-associated protein 97; hDlg; presynaptic protein SAP97; SAP-97; Synapse-associated protein 97
Gene Aliases: dJ1061C18.1.1; DLG1; DLGH1; hdlg; SAP-97; SAP97
UniProt ID: (Human) Q12959
Entrez Gene ID: (Human) 1739
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.